missing translation for 'onlineSavingsMsg'
Learn More

Human DCC Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£167.00 - £433.00

Specifications

Antigen DCC
Clone 531505
Concentration LYOPH
Dilution Immunohistochemistry 8-25 ug/mL
Applications Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18763229
GREENER_CHOICE
View Documents
R&D Systems
MAB5884-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18129560
View Documents
R&D Systems
MAB5884
100 μg
£433.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DCC Monoclonal specifically detects DCC in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

DCC
LYOPH
Immunohistochemistry
Unconjugated
Mouse
Human
P43146
1630
E. coli-derived recombinant human DCC Ser1323-Phe1447 Accession # P43146
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
531505
Immunohistochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC
DCC
IgG2b
Protein A or G purified from hybridoma culture supernatant
Detects human DCC in direct ELISAs.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.