missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DCC Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5884
This item is not returnable.
View return policy
Description
DCC Monoclonal specifically detects DCC in Human samples. It is validated for Immunohistochemistry.
Specifications
| DCC | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P43146 | |
| DCC | |
| E. coli-derived recombinant human DCC Ser1323-Phe1447 Accession # P43146 | |
| 100 μg | |
| Primary | |
| Detects human DCC in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG2b |
| Immunohistochemistry | |
| 531505 | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1630 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction