missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin SN Antibody, R&D Systems™

Goat Polyclonal Antibody
£173.00 - £415.00
Specifications
| Antigen | Cystatin SN |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795877
![]() |
R&D Systems
AF1285-SP |
25 μg |
£173.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18018821
|
R&D Systems
AF1285 |
100 μg |
£415.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin SN Polyclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Cystatin SN | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CST1, cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
| CST1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P01037 | |
| 1469 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin SN Trp21-Ser141 Accession # P01037 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Cystatin SN in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) Cystatin SA is observed, 15% cross-reactivity with rhCystatin S is observed, 5% cross-reactivity with rhCystatin C is observed, and less than 1% cross-reactivity with rhCystatin A, rhCystatin B, rhCystatin E/M, and rhCystatin F is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title