missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin SN Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1285
This item is not returnable.
View return policy
Description
Cystatin SN Polyclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Cystatin SN | |
| Polyclonal | |
| Unconjugated | |
| P01037 | |
| CST1 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin SN Trp21-Ser141 Accession # P01037 | |
| 100 μg | |
| Primary | |
| Detects human Cystatin SN in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human (rh) Cystatin SA is observed, 15% cross-reactivity with rhCystatin S is observed, 5% cross-reactivity with rhCystatin C is observed, and less than 1% cross-reactivity with rhCystatin A, rhCystatin B, rhCystatin E/M, and rhCystatin F is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| CST1, cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1469 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction