missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin SN Antibody, R&D Systems™

Mouse Monoclonal Antibody
£147.00 - £502.00
Specifications
| Antigen | Cystatin SN |
|---|---|
| Clone | 213139 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748699
![]() |
R&D Systems
MAB1285-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193331
|
R&D Systems
MAB1285 |
500 μg |
£502.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Cystatin SN | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P01037 | |
| 1469 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin SN Trp21-Ser141 (predicted) Accession # P01037 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Cystatin SN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatins A, B, C, D, S, SA, E/M, rhFetuin A, rhFetuin B, rhHPRG, or rhThrombin is observed. |
| 213139 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CST1, cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
| CST1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title