missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin SN Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1285
This item is not returnable.
View return policy
Description
Cystatin SN Monoclonal specifically detects Cystatin SN in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Cystatin SN | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P01037 | |
| CST1 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin SN Trp21-Ser141 (predicted) Accession # P01037 | |
| 500 μg | |
| Primary | |
| Detects human Cystatin SN in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatins A, B, C, D, S, SA, E/M, rhFetuin A, rhFetuin B, rhHPRG, or rhThrombin is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 213139 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CST1, cystain-SA-I, cystatin 1, cystatin SA-I, cystatin SN, cystatin-1, cystatin-SN, Salivary cystatin-SA-1, type 2 family | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1469 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction