missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cyclin B1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£178.00 - £387.00
Specifications
| Antigen | Cyclin B1 |
|---|---|
| Clone | 2061D |
| Dilution | Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunohistochemistry 3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732098
![]() |
R&D Systems
MAB60001-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766049
![]() |
R&D Systems
MAB60001-100 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry.Specifications
| Cyclin B1 | |
| Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunohistochemistry 3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Rabbit | |
| Human | |
| P14635 | |
| 891 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Detects human Cyclin B1 in direct ELISAs and Western blots. |
| 2061D | |
| Western Blot, Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| RUO | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CCNB, CCNB1, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
| E. coli-derived recombinant human Cyclin B1 Met1-Pro91 Accession # P14635 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title