missing translation for 'onlineSavingsMsg'
Learn More

Human Cyclin B1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18732098
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18732098 25 μg 25µg
18766049 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18732098 Supplier R&D Systems Supplier No. MAB60001SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cyclin B1
Applications Western Blot, Immunohistochemistry, Flow Cytometry
Classification Monoclonal
Clone 2061D
Conjugate Unconjugated
Dilution Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Immunohistochemistry 3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells
Formulation Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P14635
Gene Alias CCNB, CCNB1, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Host Species Rabbit
Immunogen E. coli-derived recombinant human Cyclin B1 Met1-Pro91 Accession # P14635
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 891
Test Specificity Detects human Cyclin B1 in direct ELISAs and Western blots.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.