missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCR6 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
£206.00 - £513.00
Specifications
| Antigen | CXCR6 |
|---|---|
| Clone | 56811 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713278
![]() |
R&D Systems
FAB699A-025 |
25 Tests |
£206.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18749086
![]() |
R&D Systems
FAB699A-100 |
100 Tests |
£513.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR6 Monoclonal specifically detects CXCR6 in Human samples. It is validated for Flow Cytometry.Specifications
| CXCR6 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Human | |
| Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
| CXCR6 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Stains human CXCR6 transfectants but not irrelevant transfectants. |
| 56811 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| O00574 | |
| 10663 | |
| NS0 mouse myeloma cell line transfected with human CXCR6 Met1-Leu342 Accession # O00574 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title