missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCR6 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems FAB699A-025
This item is not returnable.
View return policy
Description
CXCR6 Monoclonal specifically detects CXCR6 in Human samples. It is validated for Flow Cytometry.
Specifications
| CXCR6 | |
| Monoclonal | |
| APC | |
| O00574 | |
| CXCR6 | |
| NS0 mouse myeloma cell line transfected with human CXCR6 Met1-Leu342 Accession # O00574 | |
| 25 Tests | |
| 10663 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 56811 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
| Mouse | |
| Protein A or G purified from ascites | |
| Primary | |
| Stains human CXCR6 transfectants but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction