missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTRP1/C1qTNF1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
£167.00 - £451.00
Specifications
| Antigen | C1qTNF1/CTRP1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778957
![]() |
R&D Systems
AF3145-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18033252
|
R&D Systems
AF3145 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C1qTNF1/CTRP1 Polyclonal specifically detects C1qTNF1/CTRP1 in Human samples. It is validated for Western Blot.Specifications
| C1qTNF1/CTRP1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| C1q and tumor necrosis factor related protein 1, C1qTNF1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37 | |
| C1QTNF1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9BXJ1 | |
| 114897 | |
| Mouse myeloma cell line NS0-derived recombinant human CTRP1/C1qTNF1 Arg26-Pro281 Accession # Q9BXJ1 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human CTRP1/C1qTNF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CTRP4 and rhCTRP5 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title