missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTRP1/C1qTNF1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3145
This item is not returnable.
View return policy
Description
C1qTNF1/CTRP1 Polyclonal specifically detects C1qTNF1/CTRP1 in Human samples. It is validated for Western Blot.
Specifications
| C1qTNF1/CTRP1 | |
| Polyclonal | |
| Unconjugated | |
| Q9BXJ1 | |
| C1QTNF1 | |
| Mouse myeloma cell line NS0-derived recombinant human CTRP1/C1qTNF1 Arg26-Pro281 Accession # Q9BXJ1 | |
| 100 μg | |
| Primary | |
| Detects human CTRP1/C1qTNF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CTRP4 and rhCTRP5 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| C1q and tumor necrosis factor related protein 1, C1qTNF1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 114897 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction