missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRBN Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £423.00
Specifications
| Antigen | CRBN |
|---|---|
| Clone | 978015 |
| Dilution | Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18714208
![]() |
R&D Systems
MAB9574-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18707999
![]() |
R&D Systems
MAB9574-100 |
100 μg |
£423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRBN Monoclonal specifically detects CRBN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| CRBN | |
| Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| cereblon, DKFZp781K0715, MGC27358, MRT2, MRT2A, non-syndromic, autosomal recessive, 2A, protein cereblon, protein x 0001 | |
| Synthetic peptide Human CRBN Accession # Q96SW2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CRBN in immunocytochemistry and immunohistochemistry. |
| 978015 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96SW2 | |
| 51185 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title