missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRBN Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9574-100
This item is not returnable.
View return policy
Description
CRBN Monoclonal specifically detects CRBN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| CRBN | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| cereblon, DKFZp781K0715, MGC27358, MRT2, MRT2A, non-syndromic, autosomal recessive, 2A, protein cereblon, protein x 0001 | |
| Synthetic peptide Human CRBN Accession # Q96SW2 | |
| 100 μg | |
| Primary | |
| Detects human CRBN in immunocytochemistry and immunohistochemistry. | |
| Human | |
| Purified |
| Immunohistochemistry, Immunocytochemistry | |
| 978015 | |
| Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Q96SW2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51185 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction