missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Spike Protein Rabbit anti-HCoV-NL63, Clone: 279, Novus Biologicals™
Rabbit Monoclonal Antibody
£458.00
Specifications
| Antigen | Human Coronavirus Spike Protein |
|---|---|
| Clone | 279 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-NL63 samples. It is validated for ELISASpecifications
| Human Coronavirus Spike Protein | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2μm filtered solution in PBS without Preservative | |
| 3200426 | |
| IgG | |
| Protein A purified |
| 279 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| HCoV-NL63 | |
| HKU1, Spike glycoprotein | |
| This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title