missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Contactin-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£181.00 - £420.00
Specifications
| Antigen | Contactin-1 |
|---|---|
| Clone | 2332B |
| Dilution | Western Blot 2 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732557
![]() |
R&D Systems
MAB9041-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18772107
![]() |
R&D Systems
MAB9041-100 |
100 μg |
£420.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Contactin-1 Monoclonal specifically detects Contactin-1 in Human samples. It is validated for Western Blot.Specifications
| Contactin-1 | |
| Western Blot 2 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q12860 | |
| 1272.0 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2332B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CNTN1, contactin 1, contactin-1, Contactin1, F11, F3, F3cam, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
| Human embryonic kidney cell, HEK293-derived human Contactin-1, Met1-Ser993, Accession No. Q12860 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Contactin-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title