missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Contactin-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9041-SP
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Contactin-1 Monoclonal specifically detects Contactin-1 in Human samples. It is validated for Western Blot.
Specifica
| Contactin-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CNTN1, contactin 1, contactin-1, Contactin1, F11, F3, F3cam, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
| Human embryonic kidney cell, HEK293-derived human Contactin-1, Met1-Ser993, Accession No. Q12860 | |
| 25 μg | |
| 1272.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 2332B | |
| Western Blot 2 μg/mL | |
| Q12860 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Contactin-1 in direct ELISAs. | |
| Human | |
| Supernatant |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto