Learn More
Human Complement Factor H-related 1/CFHR1 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB4247
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18110611
  • £375.80 / 100µg
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
Complement Factor H-related 1/CFHR1/CFHL1 Monoclonal specifically detects Complement Factor H-related 1/CFHR1/CFHL1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
Complement Factor H-related 1/CFHR1/CFHL1
Monoclonal
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q03591
CFHR1
Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 1/CFHR1 Glu19-Ala328 Accession # Q03591
100 μg
Primary
Detects human Complement Factor H-related 1/CFHR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CFHR2 or rhCFHR5 is observed.
Human
Purified
Western Blot, Immunoprecipitation
442127
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
BTA, CFHL1FHR-1, CFHLCFHR1P, CFHR1, complement factor H-related 1, Complement Factor Hrelated 1, complement factor H-related 1 pseudogene, complement factor H-related protein 1, FHR1, FHR1CFHL1P, H factor (complement)-like 1, H factor (complement)-like 2, H factor-like protein 1, H36-1, H36-2, H-factor-like 1, HFL1H36, HFL2, MGC104329
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3078
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b