missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Factor H-related 1/CFHR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£167.00 - £416.00
Specifications
| Antigen | Complement Factor H-related 1/CFHR1/CFHL1 |
|---|---|
| Clone | 442127 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18706528
![]() |
R&D Systems
MAB4247-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18110611
|
R&D Systems
MAB4247 |
100 μg |
£416.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Factor H-related 1/CFHR1/CFHL1 Monoclonal specifically detects Complement Factor H-related 1/CFHR1/CFHL1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Complement Factor H-related 1/CFHR1/CFHL1 | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q03591 | |
| 3078 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 1/CFHR1 Glu19-Ala328 Accession # Q03591 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Complement Factor H-related 1/CFHR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CFHR2 or rhCFHR5 is observed. |
| 442127 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BTA, CFHL1FHR-1, CFHLCFHR1P, CFHR1, complement factor H-related 1, Complement Factor Hrelated 1, complement factor H-related 1 pseudogene, complement factor H-related protein 1, FHR1, FHR1CFHL1P, H factor (complement)-like 1, H factor (complement)-like 2, H factor-like protein 1, H36-1, H36-2, H-factor-like 1, HFL1H36, HFL2, MGC104329 | |
| CFHR1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title