missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Factor B Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £381.00
Specifications
| Antigen | Complement Factor B |
|---|---|
| Clone | 313011 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723318
![]() |
R&D Systems
MAB2739-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120051
|
R&D Systems
MAB2739 |
100 μg |
£381.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Factor B Monoclonal specifically detects Complement Factor B in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Complement Factor B | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P00751 | |
| 629 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor B Thr26-Leu764 Accession # P00751 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Complement Factor B in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Factor D or rhFactor I. |
| 313011 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BF, BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, CFAB, CFB, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBG, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B | |
| CFB | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title