missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Factor B Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2739
This item is not returnable.
View return policy
Description
Complement Factor B Monoclonal specifically detects Complement Factor B in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Complement Factor B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P00751 | |
| CFB | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor B Thr26-Leu764 Accession # P00751 | |
| 100 μg | |
| Primary | |
| Detects human Complement Factor B in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) Factor D or rhFactor I. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 313011 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BF, BF AHUS4, B-factor, properdin, BFD, C3 proaccelerator, C3 proactivator, C3/C5 convertase, CFAB, CFB, complement factor B, EC 3.4.21, EC 3.4.21.47, FB, FBI12, GBG, GBGCFAB, Glycine-rich beta glycoprotein, glycine-rich beta-glycoprotein, H2-Bf, PBF2, Properdin factor B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 629 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction