missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C5/C5a Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
£181.00 - £1765.00
Specifications
| Antigen | Complement Component C5/C5a |
|---|---|
| Clone | 295003 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.04-0.12 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751879
![]() |
R&D Systems
MAB2037-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18705078
![]() |
R&D Systems
MAB2037-100 |
100 μg |
£439.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18762989
![]() |
R&D Systems
MAB2037-500 |
500 μg |
£1765.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C5/C5a Monoclonal specifically detects Complement Component C5/C5a in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| Complement Component C5/C5a | |
| LYOPH | |
| Western Blot, Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 727 | |
| E. coli-derived recombinant human Complement Component C5a Thr678-Arg751 Accession # P01031 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Complement Component C5/C5a in ELISAs and Western blots. In sandwich immunoassays, detects human Complement Component C5a by itself or in the context of Complement Component C5. In Western blots, this antibody does not cross-react with recombinant human Complement Component C2, recombinant mouse (rm) Complement Component C3d, or rmComplement Component C5a. |
| 295003 | |
| Western Blot 1 ug/mL, Neutralization 0.04-0.12 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01031 | |
| C5 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title