missing translation for 'onlineSavingsMsg'
Learn More

Human Complement Component C5/C5a Antibody, R&D Systems™

Product Code. 18751879
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18751879 25 μg 25µg
18705078 100 μg 100µg
18762989 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18751879 Supplier R&D Systems Supplier No. MAB2037SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

Complement Component C5/C5a Monoclonal specifically detects Complement Component C5/C5a in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complement Component C5/C5a
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
Clone 295003
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.04-0.12 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01031
Gene Symbols C5
Host Species Mouse
Immunogen E. coli-derived recombinant human Complement Component C5a Thr678-Arg751 Accession # P01031
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Detects human Complement Component C5/C5a in ELISAs and Western blots. In sandwich immunoassays, detects human Complement Component C5a by itself or in the context of Complement Component C5. In Western blots, this antibody does not cross-react with recombinant human Complement Component C2, recombinant mouse (rm) Complement Component C3d, or rmComplement Component C5a.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.