missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C1s Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £370.00
Specifications
| Antigen | Complement Component C1s |
|---|---|
| Clone | 306904 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18720048
![]() |
R&D Systems
MAB2060-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18110981
![]() |
R&D Systems
MAB2060 |
100 μg |
£370.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Complement Component C1s Monoclonal specifically detects Complement Component C1s in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Complement Component C1s | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P09871 | |
| 716 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Component C1s Glu16-Asp688 Accession # P09871 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Complement Component C1s in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human C1r is observed. |
| 306904 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| basic proline-rich peptide IB-1, C1 esterase, C1S, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
| C1S | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title