missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Complement Component C1s Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2060
This item is not returnable.
View return policy
Description
Complement Component C1s Monoclonal specifically detects Complement Component C1s in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Complement Component C1s | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P09871 | |
| C1S | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Component C1s Glu16-Asp688 Accession # P09871 | |
| 100 μg | |
| Primary | |
| Detects human Complement Component C1s in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human C1r is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 306904 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| basic proline-rich peptide IB-1, C1 esterase, C1S, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 716 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction