missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Collagen XXV alpha 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £378.00
Specifications
| Antigen | Collagen XXV alpha 1 |
|---|---|
| Clone | 540802 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 μg/mL |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732049
![]() |
R&D Systems
MAB4317-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18149350
![]() |
R&D Systems
MAB4317 |
100 μg |
£378.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen XXV alpha 1 Monoclonal specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Immunohistochemistry.Specifications
| Collagen XXV alpha 1 | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_942014 | |
| 84570 | |
| Mouse myeloma cell line NS0-derived recombinant human Collagen XXV alpha 1 Gly189-Lys654 Accession # NP_942014 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human Collagen XXV alpha 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) COL1A1, rhCOL3A1, rhCOL4A1, rhCOL13A1, rhCOL23A1 and recombinant mouse COL13A1 is observed. |
| 540802 | |
| Immunohistochemistry 8-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, COL25A1, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
| COL25A1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title