missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Collagen XXV alpha 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4317-SP
This item is not returnable.
View return policy
Description
Collagen XXV alpha 1 Monoclonal specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Collagen XXV alpha 1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 μg/mL | |
| NP_942014 | |
| COL25A1 | |
| Mouse myeloma cell line NS0-derived recombinant human Collagen XXV alpha 1 Gly189-Lys654 Accession # NP_942014 | |
| 25 μg | |
| Primary | |
| Detects human Collagen XXV alpha 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) COL1A1, rhCOL3A1, rhCOL4A1, rhCOL13A1, rhCOL23A1 and recombinant mouse COL13A1 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 540802 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, COL25A1, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84570 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction