missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLCA1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £383.00
Specifications
| Antigen | CLCA1 |
|---|---|
| Clone | 1022517 |
| Dilution | Simple Western 20 ug/mL, Immunohistochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753557
![]() |
R&D Systems
MAB10766-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702387
![]() |
R&D Systems
MAB10766-100 |
100 μg |
£383.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLCA1 Monoclonal antibody specifically detects CLCA1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| CLCA1 | |
| Simple Western 20 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CaCC, CaCC-1, Calcium-activated chloride channel family member 1, Calcium-activated chloride channel protein 1, chloride channel accessory 1, chloride channel regulator, chloride channel regulator 1, chloride channel, calcium activated, family member 1, CLCRG1, FLJ95147, GOB5, hCaCC-1, HCLCA1 | |
| Human embryonic kidney cell HEK293-derived CLCA1, Asn22-Ser477, Accession # A8K714 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1022517 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| A8K714 | |
| 1179 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title