missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLCA1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10766-100
This item is not returnable.
View return policy
Description
CLCA1 Monoclonal antibody specifically detects CLCA1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| CLCA1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CaCC, CaCC-1, Calcium-activated chloride channel family member 1, Calcium-activated chloride channel protein 1, chloride channel accessory 1, chloride channel regulator, chloride channel regulator 1, chloride channel, calcium activated, family member 1, CLCRG1, FLJ95147, GOB5, hCaCC-1, HCLCA1 | |
| Human embryonic kidney cell HEK293-derived CLCA1, Asn22-Ser477, Accession # A8K714 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 1022517 | |
| Simple Western 20 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| A8K714 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1179 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction