missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chymase/CMA1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £388.00
Specifications
| Antigen | Chymase/CMA1/Mast Cell Chymase |
|---|---|
| Clone | 422804 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18761977
![]() |
R&D Systems
MAB40991-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792177
![]() |
R&D Systems
MAB40991-100 |
100 μg |
£388.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chymase/CMA1/Mast Cell Chymase Monoclonal antibody specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for ELISASpecifications
| Chymase/CMA1/Mast Cell Chymase | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CMA1, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCP3P, Mcpt5, MCT1, MGC119890, MGC119891 | |
| Mouse myeloma cell line NS0-derived recombinant human Chymase/CMA1, Gly20-Asn247, Accession # P23946 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 422804 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P23946 | |
| 1215 | |
| IgG3 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit