missing translation for 'onlineSavingsMsg'
Learn More

Human Chymase/CMA1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18761977
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18761977 25 μg 25µg
18792177 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18761977 Supplier R&D Systems Supplier No. MAB40991SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chymase/CMA1/Mast Cell Chymase Monoclonal antibody specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chymase/CMA1/Mast Cell Chymase
Applications ELISA
Classification Monoclonal
Clone 422804
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P23946
Gene Alias Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CMA1, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCP3P, Mcpt5, MCT1, MGC119890, MGC119891
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chymase/CMA1, Gly20-Asn247, Accession # P23946
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1215
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.