missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chymase/CMA1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £390.00
Specifications
| Antigen | Chymase/CMA1/Mast Cell Chymase |
|---|---|
| Clone | 422818 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710088
![]() |
R&D Systems
MAB4099-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18152111
|
R&D Systems
MAB4099 |
100 μg |
£390.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chymase/CMA1/Mast Cell Chymase Monoclonal specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Chymase/CMA1/Mast Cell Chymase | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P23946 | |
| 1215 | |
| Mouse myeloma cell line NS0-derived recombinant human Chymase/CMA1 Gly20-Asn247 Accession # P23946 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Chymase/CMA1 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) MCPT-1, rmMCPT-6, rmMCPT-7, or rmMCPT-11. |
| 422818 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CMA1, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCP3P, Mcpt5, MCT1, MGC119890, MGC119891 | |
| CMA1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title