missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Chymase/CMA1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4099
This item is not returnable.
View return policy
Description
Chymase/CMA1/Mast Cell Chymase Monoclonal specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Chymase/CMA1/Mast Cell Chymase | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P23946 | |
| CMA1 | |
| Mouse myeloma cell line NS0-derived recombinant human Chymase/CMA1 Gly20-Asn247 Accession # P23946 | |
| 100 μg | |
| Primary | |
| Detects human Chymase/CMA1 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) MCPT-1, rmMCPT-6, rmMCPT-7, or rmMCPT-11. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 422818 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CMA1, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCP3P, Mcpt5, MCT1, MGC119890, MGC119891 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1215 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction