missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-18 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £488.00
Specifications
| Antigen | CEACAM18 |
|---|---|
| Clone | 1057633 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736195
![]() |
R&D Systems
MAB9869-100 |
100 μg |
£488.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18785795
![]() |
R&D Systems
MAB9869-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM18 Monoclonal antibody specifically detects CEACAM18 in Human samples. It is validated for Flow CytometrySpecifications
| CEACAM18 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Carcinoembryonic Antigen-Related Cell Adhesion Molecule 18 | |
| Human embryonic kidney cell, HEK293-derived human CEACAM-18, Gln31-His317, Accession # A8MTB9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1057633 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| A8MTB9 | |
| 729767 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title