missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CEACAM-18 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9869-100
This item is not returnable.
View return policy
Description
CEACAM18 Monoclonal antibody specifically detects CEACAM18 in Human samples. It is validated for Flow Cytometry
Specifications
| CEACAM18 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Carcinoembryonic Antigen-Related Cell Adhesion Molecule 18 | |
| Human embryonic kidney cell, HEK293-derived human CEACAM-18, Gln31-His317, Accession # A8MTB9 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry | |
| 1057633 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| A8MTB9 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 729767 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction