missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDX4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£156.00 - £353.00
Specifications
| Antigen | CDX4 |
|---|---|
| Clone | 322007 |
| Concentration | LYOPH |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792909
![]() |
R&D Systems
MAB2807-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18182021
![]() |
R&D Systems
MAB2807 |
100 μg |
£353.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CDX4 Monoclonal specifically detects CDX4 in Human samples. It is validated for Immunocytochemistry.Specifications
| CDX4 | |
| LYOPH | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O14627 | |
| 1046 | |
| E. coli-derived recombinant human CDX4 Met1-Thr170 Accession # O14627 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CDX4 in direct ELISAs. |
| 322007 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| caudal type homeo box transcription factor 4, caudal type homeobox 4, caudal type homeobox transcription factor 4, Caudal-type homeobox protein 4, homeobox protein CDX-4 | |
| CDX4 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title