missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CDX4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2807
This item is not returnable.
View return policy
Description
CDX4 Monoclonal specifically detects CDX4 in Human samples. It is validated for Immunocytochemistry.
Specifications
| CDX4 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| O14627 | |
| CDX4 | |
| E. coli-derived recombinant human CDX4 Met1-Thr170 Accession # O14627 | |
| 100 μg | |
| Primary | |
| Detects human CDX4 in direct ELISAs. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 322007 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| caudal type homeo box transcription factor 4, caudal type homeobox 4, caudal type homeobox transcription factor 4, Caudal-type homeobox protein 4, homeobox protein CDX-4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1046 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction