missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD81 MAb (Clone 454729), R&D Systems™

Mouse Monoclonal Antibody
£164.00 - £410.00
Specifica
| Antigen | CD81 |
|---|---|
| Clone | 454729 |
| Dilution | Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantity | Prezzo | Quantità e disponibilità | |||||
18723017
![]() |
R&D Systems
MAB46151-SP |
25 μg |
£164.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754517
![]() |
R&D Systems
MAB46151-100 |
100 μg |
£410.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
CD81 Monoclonal specifically detects CD81 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifica
| CD81 | |
| Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD81, CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, TSPAN28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| HEK293 human embryonic kidney cell line transfected with human CD81, Accession # P60033 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD81 in direct ELISAs. |
| 454729 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Human | |
| P60033 | |
| 975 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto