missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD81 MAb (Clone 454729), R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB46151-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD81 Monoclonal specifically detects CD81 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Spezifikation
| CD81 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD81, CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, TSPAN28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| HEK293 human embryonic kidney cell line transfected with human CD81, Accession # P60033 | |
| 100 μg | |
| 975 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry, Immunocytochemistry | |
| 454729 | |
| Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL | |
| P60033 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CD81 in direct ELISAs. | |
| Human | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur