missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300c Antibody, Unconjugated, R&D Systems™, R&D Systems™

Goat Polyclonal Antibody
£156.00 - £423.00
Specifications
| Antigen | CD300c |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745087
![]() |
R&D Systems
AF3256-SP |
25 μg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18022692
![]() |
R&D Systems
AF3256 |
100 μg |
£423.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD300c Polyclonal specifically detects CD300c in Human samples. It is validated for Western Blot.Specifications
| CD300c | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CD300c antigenCMRF35 antigen, CD300c molecule, CLM6, CLM-6, CMRF35, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IGSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR | |
| CD300C | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q08708 | |
| 10871 | |
| Mouse myeloma cell line NS0-derived recombinant human CD300c/LMIR2 Met29-Arg183 Accession # Q08708 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human CD300c/LMIR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) LMIR1 is observed and less than 1% cross-reactivity with rhLMIR3, rhLMIR4, rhLMIR5, and rhLMIR6 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title