missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300c Antibody, Unconjugated, R&D Systems™, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3256
This item is not returnable.
View return policy
Description
CD300c Polyclonal specifically detects CD300c in Human samples. It is validated for Western Blot.
Specifications
| CD300c | |
| Polyclonal | |
| Unconjugated | |
| Q08708 | |
| CD300C | |
| Mouse myeloma cell line NS0-derived recombinant human CD300c/LMIR2 Met29-Arg183 Accession # Q08708 | |
| 100 μg | |
| Primary | |
| Detects human CD300c/LMIR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human (rh) LMIR1 is observed and less than 1% cross-reactivity with rhLMIR3, rhLMIR4, rhLMIR5, and rhLMIR6 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD300c antigenCMRF35 antigen, CD300c molecule, CLM6, CLM-6, CMRF35, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IGSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10871 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction