missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD200R1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£171.00 - £393.00
Specifications
| Antigen | CD200R1 |
|---|---|
| Clone | 380529 |
| Concentration | LYOPH |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18768689
![]() |
R&D Systems
MAB34141-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18109860
![]() |
R&D Systems
MAB34141 |
100 μg |
£393.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD200R1 Monoclonal specifically detects CD200R1 in Human samples. It is validated for Immunocytochemistry.Specifications
| CD200R1 | |
| LYOPH | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_620161 | |
| 131450 | |
| Mouse myeloma cell line NS0-derived recombinant human CD200 R1 Ala27-Leu266 (predicted) Accession # NP_620161 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD200 R1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD200 R1 is observed. |
| 380529 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD200 R1, CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, MOX2R, OX2RCD200 cell surface glycoprotein receptor | |
| CD200R1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title