missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD200R1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB34141
This item is not returnable.
View return policy
Description
CD200R1 Monoclonal specifically detects CD200R1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| CD200R1 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| NP_620161 | |
| CD200R1 | |
| Mouse myeloma cell line NS0-derived recombinant human CD200 R1 Ala27-Leu266 (predicted) Accession # NP_620161 | |
| 100 μg | |
| Primary | |
| Detects human CD200 R1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse CD200 R1 is observed. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 380529 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD200 R1, CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, MOX2R, OX2RCD200 cell surface glycoprotein receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 131450 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction