missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD20 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
£215.00 - £536.00
Specifications
| Antigen | CD20 |
|---|---|
| Clone | 396444 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18115441
|
R&D Systems
FAB4225V-100 |
100 Tests |
£536.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18705888
![]() |
R&D Systems
FAB4225V-025 |
25 Tests |
£215.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.Specifications
| CD20 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7 | |
| MS4A1 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 396444 | |
| Flow Cytometry | |
| Alexa Fluor 405 | |
| Mouse | |
| Human | |
| P11836 | |
| 931 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human MS4A1/CD20. Stains human MS4A1/CD20 transfectants but not irrelevant transfectants in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title