missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD20 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB4225V-100
This item is not returnable.
View return policy
Description
CD20 Monoclonal specifically detects CD20 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD20 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| B1, B-lymphocyte antigen CD20, B-lymphocyte cell-surface antigen B1, B-lymphocyte surface antigen B1, Bp35, Bp35MGC3969, CD20, CD20 antigen, CD20 receptor, CD20S7, CVID5, LEU-16, Leukocyte surface antigen Leu-16, Ly-44, Membrane-spanning 4-domains subfamily A member 1, membrane-spanning 4-domains, subfamily A, member 1, MS4A1, MS4A2, S7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 931 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 396444 | |
| Flow Cytometry 5 uL/10^6 cells | |
| P11836 | |
| MS4A1 | |
| NS0 mouse myeloma cell line transfected with human MS4A1/CD20 Met1-Pro297 Accession No. P11836 | |
| 100 Tests | |
| Primary | |
| Detects human MS4A1/CD20. Stains human MS4A1/CD20 transfectants but not irrelevant transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction