missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD163 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £402.00
Specifications
| Antigen | CD163 |
|---|---|
| Clone | R20 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18713958
![]() |
R&D Systems
MAB16072-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704969
![]() |
R&D Systems
MAB16072-100 |
100 μg |
£402.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| CD163 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Human | |
| CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Balb/c mice immunized with Synovial cells from Rheumatoid Arthritis patients | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CD163 in Flow Cytometry. |
| R20 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 9332 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title