missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD163 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB16072-100
This item is not returnable.
View return policy
Description
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD163 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CD163 in Flow Cytometry. | |
| Human | |
| Supernatant |
| Flow Cytometry, CyTOF | |
| R20 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Balb/c mice immunized with Synovial cells from Rheumatoid Arthritis patients | |
| 100 μg | |
| 9332 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction