missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL8/MCP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
£167.00 - £1540.00
Specifications
| Antigen | CCL8/MCP-2 |
|---|---|
| Clone | 35509 |
| Dilution | Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780387
![]() |
R&D Systems
MAB281-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18757669
![]() |
R&D Systems
MAB281-100 |
100 μg |
£397.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18761017
![]() |
R&D Systems
MAB281-500 |
500 μg |
£1540.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| CCL8/MCP-2 | |
| Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 6355 | |
| E. coli-derived recombinant human CCL8/MCP-2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CCL8/MCP-2 in ELISAs and Western blots. In ELISAs, this antibody shows less than 0.1% cross-reactivity with recombinant human (rh) CCL15 and rmCCL3. |
| 35509 | |
| Neutralization, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
| CCL8 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title