missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL8/MCP-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB281-100
This item is not returnable.
View return policy
Description
CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Human samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL8/MCP-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CCL8 | |
| E. coli-derived recombinant human CCL8/MCP-2 | |
| 100 μg | |
| Primary | |
| Detects human CCL8/MCP-2 in ELISAs and Western blots. In ELISAs, this antibody shows less than 0.1% cross-reactivity with recombinant human (rh) CCL15 and rmCCL3. | |
| Human | |
| Purified |
| Neutralization, ELISA | |
| 35509 | |
| Neutralization 1-4 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6355 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction