missing translation for 'onlineSavingsMsg'
Learn More

Human Cardiotrophin-1/CT-1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£157.00 - £415.00

Specifications

Antigen Cardiotrophin-1/CT-1
Clone 89221
Dilution Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18703197
GREENER_CHOICE
View Documents
R&D Systems
MAB2602-SP
25 μg
£157.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18741137
GREENER_CHOICE
View Documents
R&D Systems
MAB2602-100
100 μg
£415.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications

Specifications

Cardiotrophin-1/CT-1
Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL
Monoclonal
Mouse
Human
Q16619
1489
IgG2a
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
89221
Western Blot, ELISA, Immunohistochemistry
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1
E. coli-derived recombinant human Cardiotrophin-1 Ser2-Ala201 Accession # Q16619
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Cardiotrophin-1 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.